Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), powder

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: NSC 51986Coniferic acidNSC 2821NSC 6743203-(4-Hydroxy-3-methoxyphenyl)acrylic acidFerulic acid

F1106-25GM F1106-5GM F1106-100GM F1106-1KGBL
700014-296EA 1551 USD InStock
700014-296 700014-294 700014-300 700014-298
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), powder
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
Ferulic Acid, Powder is also known as a 3-methoxy-4-hydroxycinnamic acid and is an organic compound that is found naturally occuring in nature in seeds and cereals. It is a potent antioxidant, a protein matrix in mass spectrometry analysis and as a manufacturing precursor of vanillin.

GHS hazard : Exclamation mark


Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
Storage Temperature: Ambient
CAS Number: 1135-24-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty