Would you like to log in again?
Whatman™ Grade 114 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1114-090 1114-150 1114-090 1114-930 1114-240 1114-400 1114-150 1114-125 1114-125 1114-930 1114-240 1114-185 1114-185
70240-078PK 134.47 USD InStock
70240-078 CA70240-076 CA70240-078 CA11013-366 28464-994 70240-094 CA70240-094 70240-074 70240-084 CA28464-994 CA70240-074 70240-076 10035-254
Whatman™ Grade 114 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin.
  • Only half the thickness of Grade 113 and suitable for coarse or gelatinous precipitates
  • Smooth surface for easy recovery of precipitates
Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc. Some wet strengthened grades are available in folded (prepleated) forms.
Also available prepleated as Grade 114V.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty