Would you like to log in again?
SP Bel-Art Biohazard Disposal Bags, Plain or Printed, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F13161-0005 F13161-0009 F13162-0009 F13160-0009 F13162-0005 F13160-0005
13197-116CS 197.93 USD InStock
13197-116 47750-746 70680-346 70680-342 70680-344 13197-118
SP Bel-Art Biohazard Disposal Bags, Plain or Printed, Bel-Art Products, a part of SP
Bags Waste Disposal Bags
Great economical solution for safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes.
  • Economical
  • Available in a variety of labeling options--plain or printed
  • High temperature autoclavable at a maximum of 135 °C (275 °F)
Available plain, non-printed or with biohazard label and indicator patch. Plain, non-printed bags can also be used as conventional trash can liners.
Certifications: Meet or exceed the ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty