Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 25, 2024. During this time, our site will be not available. We appreciate your patience.
SP Bel-Art Spinpak® Magnetic Stirring Bar Six Pack Assortments, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F37160-0000 F37161-0000 F37162-0000 F37163-0000
70680-214PK 68.17 USD InStock
70680-214 70680-216 70690-154 70680-212
SP Bel-Art Spinpak® Magnetic Stirring Bar Six Pack Assortments, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
These bars are best suited for specific needs.
  • Choose from octagonal, round, micro, or color-coded bars
  • PTFE low-friction coating
  • Durable and inert
  • Convenient assortment packs suit a variety of needs
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty