Would you like to log in again?
SP Bel-Art Stainless Steel Tongs, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
H37945-0000
70690-192CS 1502.28 USD InStock
70690-192
SP Bel-Art Stainless Steel Tongs, Bel-Art Products, a part of SP
Tongs
Stainless steel, laboratory type tongs have 7.6 cm (3") tips coated with brown Teflon® FEP.
  • FEP-coated stainless steel construction
  • Coating provides anti-stick surface for easier handling
  • Suitable for handling applications of direct food contact
The coating complies with FDA regulations governing direct food contact applications and provides an anti-stick surface for easier material handling and release.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty