Would you like to log in again?
SP Bel-Art Tubing Connector Assortment, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H19570-0000
70690-194PK 94.49 USD InStock
70690-194
SP Bel-Art Tubing Connector Assortment, Bel-Art Products, a part of SP
Fittings and Connectors
This assortment of 22 stepped and straight tubing connectors comes in handy around the laboratory, and contains fittings compatible with tubing from 3.2 to 12.7 mm (⅛ to ½").
  • Chemically resistant
  • Polypropylene construction
  • Autoclavable
Chemical-resistant for use with a wide range of liquids or gases. FDA-grade polypropylene for direct contact with food, drugs, and cosmetics. Autoclavable.
Ordering information: Includes 12 Stepped Tubing Connectors and 10 Straight Tubing Connectors.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty