Would you like to log in again?
SP Bel-Art Fritware® Immersion Filters, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F13671-0000 F13670-0000
70690-276CS 1219.35 USD InStock
70690-276 70690-274
SP Bel-Art Fritware® Immersion Filters, Bel-Art Products, a part of SP
Funnels Filtration Funnels Filter Funnels
For filtration involving recovery of solids without having to transfer material to funnels.
  • HDPE construction
  • Available in a range of sizes
  • Can be precoated for fine filtration
High-density polyethylene plastic coarse porosity disc (90-130 micron) and body. Use directly or precoat for fine filtration. Larger filter has a serrated tubing connector.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty