Would you like to log in again?
I-CHEM™ Clean Snap Vials with 0.125" Septa, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SS236-0040 SS146-0040 SS246-0040 SS346-0040 SS336-0040 SS136-0040
71000-088CS 327.32 USD InStock
71000-088 71000-082 71000-090 71000-080 71000-086 71000-084
I-CHEM™ Clean Snap Vials with 0.125" Septa, Thermo Scientific
Vials
Protect samples from contamination during storage or transport with Thermo Scientific™ Clean snap clear vials.
An innovative snap-off cap reduces the chance of volatile organic contaminants in the environment from passing through the septum while also keeping the septum clean and free from foreign particles during handling. The clean snap cap easily peals or snaps away to provide access to the septum at the time of analysis.
Certifications: Certified to meet or exceed EPA performance-based standards for volatile organic analysis.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty