Would you like to log in again?
Y-Type Connectors, Ace Glass Incorporated
Supplier: Ace Glass
12514-04 12514-12 12514-08 12514-10 12514-14
71000-232CS 567.06 USD InStock
71000-232 71000-230 71000-226 71000-224 71000-228
Y-Type Connectors, Ace Glass Incorporated
Fittings and Connectors
Versatile "Y" connectors are fabricated from polypropylene
They are autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty