Would you like to log in again?
Filling Bell Aseptic, Ace Glass Incorporated
Supplier: Ace Glass
3868-02 3868-04 3868-10 3868-07 3868-05
71000-320CS 77.9 USD InStock
71000-320 71000-314 71000-312 71000-318 71000-316
Filling Bell Aseptic, Ace Glass Incorporated
Bottles Cell Culture Bottles
This apparatus is used for aseptic filling of vessels.
  • Fabricated from borosilicate glass
Rubber tubing is attached to the top hose connection, the receiving vessel is placed inside the bell.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty