Would you like to log in again?
Stainless Steel Closures, Ace Glass Incorporated
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
3918-50 3918-53 3918-59 3918-51 3918-56 3918-52 3918-54 3918-61 3918-55 3918-58 3918-57
71000-332CS 765.27 USD InStock
71000-332 71000-340 71000-330 71000-324 71000-336 10098-726 71000-322 71000-334 71000-328 71000-326 71000-338
Stainless Steel Closures, Ace Glass Incorporated
Closures Test/Sample Tube Closures
These closures are designed for use with culture tubes and flasks and are available with or without gripping finger indents
Closures are made of 305 Stainless Steel and are fully autoclavable.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty