Would you like to log in again?
Flask, Culture, Fernbach, Ace Glass Incorporated
Supplier: Ace Glass
3874-18 3874-30
71000-342CS 695 USD InStock
71000-342 71000-348
Flask, Culture, Fernbach, Ace Glass Incorporated
Flasks Cell Culture Flasks
This wide mouth Fernbach flask is designed for culturing organisms requiring a large surface-area-to-volume ratio
Triple baffles are on the bottom edges to achieve maximal oxygen transfer to the culture medium. Beaded neck is 63 mm I.D,. and it accepts cotton plugs or rubber stoppers. Fabricated from borosilicate glass.
Ordering information: Rubber stoppers and/or cotton plugs NOT included. Caution: Do not place on direct heat source.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty