Would you like to log in again?
Flask, Fernbach, Baffled, Culture, Ace Glass Incorporated
Supplier: Ace Glass
3875-31 3875-20
71000-344CS 1042.95 USD InStock
71000-344 71000-350
Flask, Fernbach, Baffled, Culture, Ace Glass Incorporated
Flasks Cell Culture Flasks
This Fernbach culture flask has a long-style 38mm neck for plastic or stainless steel closures
Triple baffles are located at the center of the bottom to achieve maximal oxygen transfer to culture medium. Fabricated from borosilicate glass.
Ordering information: Closure NOT included. Caution: Do not place on direct heat source.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty