Would you like to log in again?
Nephelo Culture Flasks with Screw Caps, Ace Glass Incorporated
Supplier: Ace Glass
3910-03 3910-35 3910-07 3910-31 3910-22 3910-26 3910-24 3910-33
71000-386CS 592.98 USD InStock
71000-386 71000-354 71000-362 71000-374 71000-368 71000-358 71000-356 71000-382
Nephelo Culture Flasks with Screw Caps, Ace Glass Incorporated
Flasks Cell Culture Flasks
Nephelo culture flask with side port and angled side arm cuvette for standard nephelometers and spectrophotometers
Supplied with a non-toxic, rubber-lined 38-430 phenolic top cap and a 18-415 clean-out port cap. Ideal for the growing and mixing of aerobic and anaerobic cultures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty