Would you like to log in again?
Trypsinizing Flask, Baffled, Ace Glass Incorporated
Supplier: Ace Glass
3897-14 3897-25 3897-04 3897-21 3897-08 3897-12 3897-19
71000-496EA 32.42 USD InStock
71000-496 71000-498 71000-510 71000-504 71000-502 71000-492 71000-506
Trypsinizing Flask, Baffled, Ace Glass Incorporated
Flasks Trypsinization Flasks
Trypsinizing flask with beaded neck and long side baffles
Ideal for converting homogenous tissue samples into cell suspensions by digestion of connective tissue. Heavy beaded neck accepts either cotton plugs or stoppers. The extra-deep baffles enhance vigorous agitation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty