Would you like to log in again?
Tissue Homogenizer, Potter-Elehjem, Ace Glass Incorporated
Supplier: Ace Glass
8355-05 8355-10 8355-30 8355-02 8355-15 8355-55
71000-530CS 681.59 USD InStock
71000-530 71000-522 71000-532 71000-526 71000-524 71000-528
Tissue Homogenizer, Potter-Elehjem, Ace Glass Incorporated
Homogenizers Tissue Grinders
When using this Potter-Elehjem tissue homogenizer, homogenization occurs as the sample and buffer are forced through the cylindrical portion of the mortar as the pestle is rotated downward
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
The mortar is not ground, and the pestle contact surface is PTFE, threaded onto a stainless steel shaft. Tube fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty