Would you like to log in again?
Wide Mouth Glass Packer Bottles, Amber, C&G Containers
Supplier: C&G Containers
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
LAP002900000 LAP042200010 LAP016900000 LAP004100010 LAP083200000 LAP083900000 LAP008900000 LAP002200002 LAP008200110 LAP032210010 LAP042200000 LAP004100000 LAP008100010 LAP004900000 LAP042900000 LAP008210010 LAP032230010 LAP002308100 LAP008230010 LAP004900002 LAP008100100 LAP032900000 LAP032100000 LAP032200000 LAP016100000 LAP002400010 LAP083200010 LAP008900100 LAP008200010 LAP002400110 LAP004200010 LAP016200010 LAP008308100 LAP004200100
71000-672CS 111.1 USD InStock
71000-672 71000-650 71000-676 71000-654 71000-674 71000-652 71001-758 30617-854 89137-856 89137-858 10496-900 71000-668 30621-146 71000-684 71000-662 71000-660 71000-682 30621-148 71000-666 71000-644 71000-664 71000-642 10496-902 71000-680 30621-150 71001-632 71000-658 71000-636 71000-678 71000-656 71000-634 30621-152 71000-638 30621-154
Wide Mouth Glass Packer Bottles, Amber, C&G Containers
Jars General Use Jars
These glass jars are used primarily for the collection of liquid media, soil, sediments, and slurry media for testing Semi-volatiles (Light Sensitive compounds), Pesticides, PCBs, Diesel Range Organics, TPH, Metals, and Phenolics
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash). Some items are Nitrogen-purged, others are chemically preserved.
Secondary uses include containment, bench vessels, and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity "contain" volumes may vary.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty