Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

OSOM® Trichomonas Rapid Test Kit, Sekisui Diagnostics

Supplier: Sekisui Diagnostics

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

OSOM®
181 181 182 182
71001-054EA 50.99 USD InStock
CA71001-052 71001-054 CA71001-054 71001-052
OSOM® Trichomonas Rapid Test Kit, Sekisui Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Trichomonas Rapid Test is an immunochromatographic assay designed to detect Trichomonas Vaginalis antigens directly from vaginal swabs, so a speculum may not be required

Control kit (sold separately) includes additional controls for the trichomonas test.

Detects the antigen, removing the uncertainty with live organism instability. Sensitivity is 83% versus culture. Studies show wet mount sensitivity is <60%. Kits should be stored at room temperature to maintain a 16-month shelf life.

Certifications: CLIA-waived. CE marked.

Ordering information: Test kit includes controls. Control kit includes 10 positive control swabs.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty