Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

OSOM® BVBlue® Test Kit, Sekisui Diagnostics

Supplier: Sekisui Diagnostics

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

OSOM® BVBlue® OSOM®
184 183
71001-056CS 2663.1 USD InStock
71001-056 71001-058
OSOM® BVBlue® Test Kit, Sekisui Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
Rapid test for the diagnosis of bacterial vaginosis. Detects elevated vaginal fluid sialidase activity, an enzyme produced by bacterial pathogens associated with bacterial vaginosis including Gardnerella, Bacteroides, Prevotella and Mobiluncus.

  • 92.8% sensitive, 98% specific versus Gram Stain
  • 1 minute hands-on-time; 10 minute read time
  • Instant color change provides clear easy-to-read results.
  • CPT Code: 87905QW

Detects all four major pathogens which cause BV. Test uses a vaginal swab; a speculum may not be required. Refrigerated storage at 2–8°C (35.6–46.4°F).

Certifications: CLIA-waived.

Ordering information: Control kit (sold separately) includes one positive and one negative control.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty