Would you like to log in again?
Plastic Packers, Wide Mouth, HDPE, Natural, C&G Containers
Supplier: C&G Containers
LPW004200011 LPW008900000 LPW064200010 LPW008200012 LPW008230012 LPW008220012 LPW128200010 LPN128900002 LPW128900000 LPW016200010 LPW002200010 LPW002200011 LPW032230013 LPW008200010 LPW016220012 LPW004900000 LPW032200010 LPW032900003 LPW016230012 LPW032900000 LPW016900002 LPW008900002 LPW004200010 LPW002900000 LPW032200013 LPW066200010 LPW064900004 LPW016200012
71001-116CS 167.7 USD InStock
71001-116 71001-138 71001-118 71001-142 71001-164 71001-122 71001-144 71001-166 71001-124 71001-146 71001-148 71001-160 71001-162 71001-140 71001-128 71001-106 71001-108 71001-130 71001-174 71001-152 71001-010 71001-132 71001-110 71001-134 71001-136 71001-158 71001-114 71001-170
Plastic Packers, Wide Mouth, HDPE, Natural, C&G Containers
Bottles Water Sampling Bottles
These plastic bottles are used for the collection of aqueous media for testing metals and a variety of inorganic parameters, including alkalinity, anions, acidity, cyanide, sulfate, and hardness.
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash). Some items are chemically preserved.
Not all analytical parameters are included on the Certificate of Analysis. Please refer to the Certificate of Analysis for certified analytes and corresponding reporting limits. Secondary uses for these containers include containment and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty