Would you like to log in again?
Ace-Safe PTFE Tubing Connectors, Ace Glass Incorporated
Supplier: Ace Glass
5858-212 5858-210 5858-12 5858-03 5858-07 5858-05 5858-10 5858-14
76318-506EA 48.18 USD InStock
76318-506 76318-508 71001-520 71001-510 71001-512 71001-514 71001-516 71001-518
Ace-Safe PTFE Tubing Connectors, Ace Glass Incorporated
Fittings and Connectors
These complete PTFE tubing connectors and PTFE bushings make an easy and safe connect or disconnect of coolant or steam tubing to jacketed reaction flasks, condensers, or any vessel utilizing the Ace-Thred as the connection port.
Ordering information: Also available in polypropylene. Contact your Avantor representative for more information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty