Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Flush Seal Bottom Drain Valve Assembly with Pin, Ace Glass Incorporated
Supplier: Ace Glass
6482-20
71001-526EA 2257.26 USD InStock
71001-526
Flush Seal Bottom Drain Valve Assembly with Pin, Ace Glass Incorporated
Valves Line Valves
Redesigned flush-seal type valve with the top of the valve having a 1-1/2" x 3/8" O.D
pin that extends upward. The pin fits into the receptacle on the bottom of certain ACE agitators. This assembly is designed to reduce shaft wobble or “flexing” on the larger reactors. Top opening is 2-inch beaded pipe and the side drain port is 1-inch beaded pipe. Uses clamp from product ID#89064-666.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty