Would you like to log in again?
Conical Flange, 4 Neck Reaction Head, Ace Glass Incorporated
Supplier: Ace Glass
6499-14 6499-10 6499-18
71001-530EA 620.92 USD InStock
71001-530 71001-532 71001-528
Conical Flange, 4 Neck Reaction Head, Ace Glass Incorporated
Reaction Vessels Reaction Vessels, Flask Shape
Conical flange with 4" (100 mm) opening
All the side necks are 24/40. Use clamp to secure head. For use with certain ACE reaction flasks. Fabricated from borosilicate glass.
One standard taper center joint and three standard taper side necks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty