Would you like to log in again?
Multi-Paddle Agitators, PTFE, 19 mm, Ace Glass Incorporated
Supplier: Ace Glass
8100-19 8100-09
71001-572EA 798.87 USD InStock
71001-572 71001-574
Multi-Paddle Agitators, PTFE, 19 mm, Ace Glass Incorporated
Stirrers Stirring Shafts
This is a PTFE, large-scale, multi-paddle agitator design to accept pinned bottom valves
The pin helps reduce wobble or flexing at higher rpm. 19mm agitators will fit drilled type or knob type shafts.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty