Would you like to log in again?
PYREX® 3-Hole Mobile Phase Delivery Cap, Corning
Supplier: Corning
PYREX®
1395-3H
71001-620CS 329.76 USD InStock
71001-620
PYREX® 3-Hole Mobile Phase Delivery Cap, Corning
Closures Safety Caps and Safety Waste Caps
The delivery cap top has three holes that have 1/4" - 28 threads with 1/8" O.D
For use with PYREX® HPLC Reservoirs, Single Cavity.
PTFE tubing. The body is glass-filled PTFE (Polytetrafluoroethylene). PET (Polyethylene terephthalate) screw cap has a GL-45 thread. The cap includes three 1/4" - 28 ETFE (Ethylene tetrafluoroethylene) plugs and a Viton® O-ring for sealing to the reservoir. Integral pressure relief valve relieves reservoir pressure at 10 psi.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty