Would you like to log in again?
PolarSafe™ White and Colored Cardboard Freezer Storage Boxes And Dividers, Argos Technologies
Supplier: Argos Technologies
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
PolarSafe™
04395-41 04395-40 04395-35 06755-71 06755-81 06755-74 06755-83 06755-84 R3016A 06755-86 06755-73 06755-76 04395-37 04395-39 06755-82 06755-79 06755-80 04395-34 06755-72 06755-75 04395-38 06755-78 06755-85 04395-36
10030-118CS 408.08 USD InStock
10030-118 10770-466 10436-316 71001-644 10436-314 10030-120 10436-318 89128-176 89128-198 89128-186 89128-200 89128-178 89128-188 89128-202 71001-640 10436-322 71001-642 10436-320 89128-190 89128-192 89128-194 89128-183 89128-196 89128-184
PolarSafe™ White and Colored Cardboard Freezer Storage Boxes And Dividers, Argos Technologies
Boxes Cryoboxes
PolarSafe™ cryo/freezer boxes and dividers are made of sturdy white cardboard, with a moisture-repellent outer coating to protect from the effects of liquid nitrogen or condensation from refrigerated equipment
The 81 and 100 place boxes have a moisture repellent lining on the inside of the box as well as on the dividers. In addition, both boxes have been printed with a unique numerical grid to keep your samples organized.
Two specialty boxes for PCR are available. The first is 5 x 5 x 1 and will accommodate a 196 place cardboard divider. The 1" height prevents the tubes from dropping down into the box. The second is a 64 place PCR box that will hold 8 PCR strips. This can also be used as a PCR rack to transport tubes to the Thermocycler.
The colored boxes are available in 5 bright colors. The color is only located around the outer sides of the boxes leaving the top white for labeling or for the researchers identification.
Choice of boxes and dividers to accommodate tubes from 0.5 - 50 mL.
Boxes are sized to fit all standard stainless-steel and polycarbonate storage systems. Small holes on the bottom allow for safe drainage of liquid nitrogen or water. For use in mechanical freezers and liquid nitrogen tanks at temperature to -196°C.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty