Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Open Ring Supports with Extension Arms, Ace Glass Incorporated
Supplier: Ace Glass
11177-17 11177-19 11177-21 11177-13
71001-718EA 194.72 USD InStock
71001-718 10099-140 71001-714 71001-716
Open Ring Supports with Extension Arms, Ace Glass Incorporated
Support Rings
Open ring supports with long extension arms for supporting glassware from the bottom, such as separatory funnels and powder funnels
PVC coated ring also protects glass from scratching. Long extension arms allow for easier connection to labframes or stands.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty