Would you like to log in again?
Drop In Sink, Self Rimming, Two Bowl, Eagle MHC
Supplier: Eagle MHC
SR18-24-13.5-2 SR16-19-13.5-2 SR10-14-9.5-2 SR22-22-13.5-2 SR24-24-13.5-2 SR16-19-8-2 SR12-14-9.5-2 SR14-16-9.5-2
71001-916EA 3683.31 USD InStock
71001-916 71001-906 71001-918 71001-908 71001-920 71001-910 71001-912 71001-914
Drop In Sink, Self Rimming, Two Bowl, Eagle MHC
Furniture Casework
Heavy gauge type 304 series stainless steel construction.
  • Faucet holes punched on 4" centers
  • 3 to ½" drain hole
Two compartment. Includes mounting hardware, faucet with 12" spout and basket drain.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty