Would you like to log in again?
TOS-Propionate Agar (Base)
Supplier: EMD Millipore
1.00043.0500 1.00043.0500
71002-338EA 805.88 USD InStock
71002-338 CA1.00043.0500
TOS-Propionate Agar (Base)
Media for Microbiology Dehydrated Media for Microbiology
For the enumeration of Bifidobacteria in dairy products. 62.5g in 950mL of purified water.
Contents: peptone from casein 10.0; Yeast extract 1.0; potassium di-hydrogen phosphate 3.0; Di-potassium hydrogen phosphate 4.8; Ammonium sulfate 3.0; Magnesium sulfate heptahydrate 0.2; L-cysteine monochloride 0.5; Sodium Propionate 15.0; Galactooligosaccharide(TOS) 10.0; Agar Agar 15.0.
For the enumeration of Bifidobacteria in dairy products
62.5g in 950mL of purified water
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty