Would you like to log in again?
Orion™ Conductivity Standards, Solutions, and Accessories, Thermo Scientific
Supplier: Thermo Scientific
Orion™
011006 11007 011005 011005 11008 990106
52458-147CS 181.94 USD InStock
52458-147 CA011005 52458-149 71002-397 52458-145 23227-504
Orion™ Conductivity Standards, Solutions, and Accessories, Thermo Scientific
Standards Conductivity Standards
Perform virtually any electrochemistry analysis with Orion™ Conductivity Standards and Solutions
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards
Conductivity standard temperature tables are programmed into Orion™ meter software for use with the automatic calibration feature.
Choose from a complete selection of conductivity accessories including conductivity standards and TDS (total dissolved solids) standards. All conductivity standards are traceable to NIST standard reference materials.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty