Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Ethanol ≥70%, KOPTEC USP, Biotechnology Grade (140 Proof)

Supplier: Decon Labs
Danger
Synonyms: Ethyl alcohol

V1401 V1455TP V1405HCTP V1405HC V1455MTP V1401TP
71001-652CS 171.83 USD InStock
71001-652 89234-862 71001-654 71002-332 89234-860 71002-508
Ethanol ≥70%, KOPTEC USP, Biotechnology Grade (140 Proof)
Ethanol
140 proof pure (undenatured) ethanol (70%) USP. Used for research, healthcare, industrial, and production applications as a cleaner, solvent, and reagent.

  • Total alcohol: 70%
  • Ethyl alcohol: 70%
  • Denaturant: none

Ordering information: Tax-paid ethanol prices include the federal excise tax on pure ethanol. To purchase tax-free ethanol, customers must possess an Industrial Alcohol User Permit from TTB, specifically marked for the withdrawal and use of alcohol free of tax.


Formula: C₂H₅OH
MW: 46.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty