Would you like to log in again?
GST Buffer Kit, Cytiva
Supplier: Cytiva
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
28-9523-61 28952361
71002-602KT 629.06 USD InStock
CA71002-602 71002-602
GST Buffer Kit, Cytiva
Buffers Biological Buffers
GST Buffer Kit contains carefully prepared buffer concentrates for binding, washing, and elution of GST-tagged proteins according to recommended protocols.
  • Premade stock solutions of buffers optimized for GST-tagged protein purification
  • Sufficient reagents are supplied to purify up to 20 mg of GST-tagged protein
GST Buffer Kit eliminates time-consuming buffer preparation and allows fast, reproducible, and convenient purification work. GST Buffer Kit can be used in combination with a number of column types: GST SpinTrap, GST MultiTrap 96-well plates, GST GraviTrap, GSTrap FF, GSTrap 4B and GSTrap HP.
Delivery information: Includes 50 ml 10 × PBS stock solution pH 7.3 (1.4 M NaCl, 27 mM KCl, 101 mM Na2HPO4, 18 mM KH2PO4, pH 7.4); Reduced Glutathione 0.154 g. Addition of 50 ml of glutathione dilution buffer results in a solution of 10 mM glutathione, 50 mM Tris-HCl (pH 8.0); 50 ml dilution buffer: 50 mM Tris-HCl (pH 8.0).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty