Would you like to log in again?
HiLoad Superdex Columns, 200 PG, Cytiva
Supplier: Cytiva
HiLoad™
28-9893-36 28989335 28989336 28-9893-35
71002-672EA 6449.57 USD InStock
CA71002-672 71002-672 CA71002-670 71002-670
HiLoad Superdex Columns, 200 PG, Cytiva
Chromatography Columns
Superdex prep grade (pg) is produced by covalent bonding of dextran to highly cross-linked agarose.The separation properties of these media are determined by the dextran component.
  • High resolution with short run times and good recovery
  • High capacity at high flow rates
  • Separations can be scaled up to production levels
  • Prepacked for convenience and reproducibility
  • 1/16" fittings for convenient connections to ÄKTA design systems
Steep selectivity curves give excellent resolving power for peptides and proteins in the molecular weight ranges, Mr <10000 (Superdex 30 pg), Mr 3000 to 70000 (Superdex 75 pg), and Mr 10000 600000 (Superdex 200 pg). The medium combines high mechanical strength with high hydrophilicity, allowing high flow rates, and minimal nonspecific interactions.
Delivery information: Includes dismantling tool, support screen, net ring (10 μm), O-ring, stop plugs and instructions.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty