Would you like to log in again?
RedDot™1 Far-Red Nuclear Stain, 200X in Water, Biotium
Supplier: Biotium
RedDot™
40060-T 40060 40060-1
71002-678EA 313.51 USD InStock
71002-678 71002-682 71002-680
RedDot™1 Far-Red Nuclear Stain, 200X in Water, Biotium
Assays Cellular Assays
RedDot™1 is a far-red cell membrane-permeable nuclear dye similar to Draq5™. The dye is ideal for specifically staining the nuclei of live cells.
  • Ideal for specifically staining the nuclei of live cells for short term imaging
  • Highly thermostable and photostable, for convenient handling and demanding imaging applications
  • Can be used for DNA content analysis by flow cytometry like Vybrant™ DyeCycle™ Ruby
  • Can be used for normalization of In Cell Western® in fixed cells, similar to Draq5™/Sapphire®700
  • λEx/λEm = 662/694 nm (with DNA), for detection in the Cy®5 channel
RedDot™1 can be excited at a wide range of wavelengths between 488 nm and 647 nm, including the 488 nm laser for flow cytometry, and can be used for DNA content analysis by flow cytometry. While DAPI and Hoechst dyes show a strong preference for A-T rich regions, RedDot™1 is relatively insensitive to sequence base composition.
Like Draq5®, RedDot™1 is toxic to cells within a few hours, and is recommended for short term staining experiments. For a low-toxicity far-red nuclear stain for live cells, see our NucSpot™ 650 dye.
RedDot™1 cannot be used for nuclear-specific staining in fixed cells. Please see RedDot™2 (cat# 40061), a spectrally similar dye designed for specific nuclear staining of fixed and permeabilized cells or selective nuclear staining of dead cells.
Please note: far-red dyes like RedDot™1 are not visible to the human eye, but must be imaged with a CCD camera or by confocal microscopy.
Cy dye is a registered trademark of GE Healthcare. Draq5 is a trademark of BioStatus Ltd. In Cell Western and Sapphire are registered trademarks of LI-COR Inc. Vybrant and DyeCycle are trademarks of Thermo Fisher Scientific.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty