Would you like to log in again?
Glutathione Resin for GST Purification, G-Biosciences
Supplier: G-Biosciences
786-280 786-311 786-310 786-312 786-280
71003-128EA 868.1 USD InStock
CA71003-128 CA82022-712 71003-128 82022-712 CA71003-130 71003-132 CA71003-132 71003-130
Glutathione Resin for GST Purification, G-Biosciences
Protein Linking and Labeling Systems
Glutathione Resin is designed for the affinity purification of proteins with a glutathione S-transferase (GST) tag. The resin consists of reduced glutathione (GSH) covalently coupled to 6% cross-linked agarose, via a 10-carbon spacer arm. The resin has a binding capacity of >40mg GST/ml resin.
  • For the purification of GST proteins
  • High capacity (>40mg/ml)
  • Bead size: 50-160μm
  • Bead structure: 6% cross-linked agarose
  • 10 carbon spacer arm
The resin, columns and binding/wash and elution buffers are available separately.
The spin columns are supplied with a resin bed volume of 0.2, 1 and 3ml with total column volumes of 1, 8 and 22ml respectively. Columns can be used as a spin format of gravity flow columns.
Also available in 96-well spin plate formats for processing up to 96 samples.
G-Biosciences' Glutathione Resin is supplied as slurry in 20% ethanol. It is available as a resin alone or supplied in a kit format.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty