Would you like to log in again?
Jacalin for IgA Purification, G-Biosciences
Supplier: G-Biosciences
786-473 786-167 786-473 786-167
71003-088EA 392.5 USD InStock
CA71003-186 71003-088 CA71003-088 71003-186
Jacalin for IgA Purification, G-Biosciences
Protein Purification Protein Purification Resins
G-Biosciences' Immobilized Jacalin is designed for preparing Human IgA free of contaminating IgG
Jacalin, or Artocarpus integrifolia lectin, is a tetrameric two-chain lectin with a molecular weight of 66 kDa. Jacalin is a α-D-galactose binding lectin purified from jack-fruit (Artocarpus integrifolia) seeds.
G-Biosciences' Jacalin is also available in lyophilized form. Applications include isolating IgA from human serum and colostrums, isolating human plasma glycoproteins and histochemistry. Jacalin also binds IgD.
The binding capacity of G-Biosciences' Immobilized Jacalin is 1-3 mg human IgA per mL resin.
Ordering information: Accessories available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty