Would you like to log in again?
Gram Negative Lysis Buffer, G-Biosciences
Supplier: G-Biosciences
786-580 786-579 786-567 786-566
71003-190EA 220 USD InStock
71003-190 71003-192 CA71003-190 CA89167-786 CA89167-784 89167-786 89167-784 CA71003-192
Gram Negative Lysis Buffer, G-Biosciences
G-Biosciences' Gram-negative Lysis Buffer is an extraction buffer for soluble proteins from Gram-negative bacteria
Depending on the application, additional agents such as reducing agents, chelating agent, nuclease and protease inhibitors cocktail may be added into Gram-negative lysis buffer. This buffer has been tested for use with several widely used bacteria including E. coli strains.
It is a proprietary improvement on the lysozyme based lysis in combination with various salts and agents, which allows extraction of soluble proteins.
Ordering information: Accessories available.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty