Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium carbonate, anhydrous

Supplier: G-Biosciences
Warning
Synonyms: Na-XCarbonic acid sodium saltDisodium carbonateSodaBisodium carbonateCarbonic acid, disodium saltSoda ash

RC-126 RC-125 RC-125 RC-126
71003-526EA 141.07 USD InStock
CA71003-524 71003-526 CA71003-526 71003-524
Sodium carbonate, anhydrous
Sodium carbonate


Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/89/16640589.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty