Would you like to log in again?
RODAC™ Contact Plates, TSA with Lecithin and Polysorbate 80, BD Diagnostics
Supplier: BD Diagnostics
215277 299896 222207 215276 215285 215278 222207 215275 292335 215278 292335 215275 221238 215276 215285
71003-768CS 627.49 USD InStock
CA71003-766 71003-768 CA71003-768 71003-786 CA71003-772 CA71003-786 71003-766 CA90005-296 90001-274 CA90004-226 90001-614 CA71003-770 71003-772 90004-226 CA-90001-614
RODAC™ Contact Plates, TSA with Lecithin and Polysorbate 80, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
BD RODAC™ contact plates are available with or without the BD RODAC™ Snap lid. The BD RODAC™ Snap Lid was designed by environmental monitoring professionals to provide additional security during sampling and transport from a controlled environment to the laboratory.
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty