Would you like to log in again?
Contact Plates , SDA with Lecithin and Polysorbate 80, BD Diagnostics
Supplier: BD
292337 215282 292351 292653 215270 221233 215282 292337 292351 221233 292653 215270
90001-268PK 46.53 USD InStock
90001-268 71003-756 CA90001-268 CA71003-756 CA71003-780 90007-922 90004-231 CA90004-230 CA90007-922 90007-764 71003-780 CA90007-764
Contact Plates , SDA with Lecithin and Polysorbate 80, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
The BD RODAC™ Snap Lid was designed by environmental monitoring professionals to provide additional security during sampling and transport from a controlled environment to the laboratory.
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty