Would you like to log in again?
Alpha-Bungarotoxin, CF® Dye and Other Conjugates, Biotium
Supplier: Biotium
00010-1 00026 00004 00003 00015 00007-100UG 00006 00003-100UG 00005-100UG 00012 00005 00002-100UG 00002 00018-100UG 00016 00011 00006-100UG 00013 00004-100UG 00018 00017 00007 00014 00026-100UG 00009-100UG 00009
76221-716EA 149.38 USD InStock
76221-716 89138-088 76221-718 89138-086 89138-084 89138-082 71004-296 71004-006 76221-708 76221-704 10018-138 76221-706 89138-078 89138-096 89138-074 89138-072 89138-094 89138-092 89138-070 76423-664 89138-090 71004-058 76221-712 71003-982 76221-714 76221-710
Alpha-Bungarotoxin, CF® Dye and Other Conjugates, Biotium
Protein Linking and Labeling Systems
Conjugates of alpha-bungarotoxin labeled with a selection of our CF® dyes and other labels. Labeled alpha-bungarotoxin conjugates can be used for staining nicotinic acetylcholine receptors at neuromuscular junctions in tissue sections.
  • Choose from nine bright and stable CF® Dye colors or other labels
  • Dye options for super-resolution and 2-photon imaging
A variety of alpha-bungarotoxin CF® Dye conjugates offers advantages in brightness, photostability, and/or signal-to-noise ratio compared to other fluorescent dyes. Also available alpha-bungarotoxin conjugates with biotin, fluorescein (FITC), tetramethylrhodamine (TRITC), or sulforhodamine-101 (Texas Red®) and unconjugated alpha-bungarotoxin.
CF® dyes are covered by pending U.S. and international patents. CF® is a trademark of Biotium, Inc. Texas Red® is a registered trademark of Molecular Probes, Inc.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty