Would you like to log in again?
CF™ Dye Bovine Serum Albumin Conjugates, Biotium
Supplier: Biotium
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
20289 20291 20292 20290
71004-204EA 296.02 USD InStock
71004-204 71004-202 71004-208 71004-210
CF™ Dye Bovine Serum Albumin Conjugates, Biotium
Protein Linking and Labeling Systems
Fluorescent bovine serum albumin (BSA) conjugates are commonly used as tracers in applications where physical dimensions are important
CF dye technology is covered by pending U.S. and international patents.
The conjugates can also be used for quantitative studies of electroporation, measurement of plasma volume in rats, and endocytosis studies. CF™ dye BSA conjugates are labeled with Biotium’s next-generation fluorescent CF™ dyes, which offer advantages in brightness and photostability compared to other fluorescent dyes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty