Would you like to log in again?
V.C.N. Supplement, HiMedia Laboratories
Supplier: HiMedia Laboratories
FD023-5X5VL FD023-5X5VL FD023-5VL FD023-5VL
71005-610PK 448.67 USD InStock
71005-610 CA71005-610 71005-608 CA71005-608
V.C.N. Supplement, HiMedia Laboratories
Media for Microbiology Additives and Supplements for Microbiology
An antibiotic supplement, recommended for the selective isolation of N. gonorrhoeae and N. meningitidis.
  • Store at 2 to 8 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty