Would you like to log in again?
Cetrinix Supplement, HiMedia Laboratories
Supplier: HiMedia Laboratories
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FD029-5VL FD029-5X5VL FD029-5VL FD029-5X5VL
71005-624PK 100.06 USD InStock
71005-624 CA71005-624 71005-626 CA71005-626
Cetrinix Supplement, HiMedia Laboratories
Media for Microbiology Additives and Supplements for Microbiology
An antibiotic supplement recommended for the selective isolation of Pseudomonas species.
  • Store at 2 to 8 °C
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty