Would you like to log in again?
Rosolic Acid, HiMedia
Supplier: HiMedia Laboratories
FD058-5VL FD058-5VL
71005-688PK 72.6 USD InStock
71005-688 CA71005-688
Rosolic Acid, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Rosolic acid solution is recommended for selective isolation of coliform bacteria using membrane filter technique at higher temperature (44.5 ˚C).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty