Would you like to log in again?
Tellurite-Cefixime Supplement, HiMedia Laboratories
Supplier: HiMedia Laboratories
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
FD147-5VL FD147-5VL
71005-838PK 64.1 USD InStock
71005-838 CA71005-838
Tellurite-Cefixime Supplement, HiMedia Laboratories
Media for Microbiology Additives and Supplements for Microbiology
These supplements are ideal for isolation of Escherichia coli O157:H7
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty