Would you like to log in again?
TALON® Superflow™ Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Talon®
28957502 28957499 28-9574-99 28-9575-02
71006-008EA 950.76 USD InStock
CA71006-008 CA71006-006 71006-008 71006-006
TALON® Superflow™ Affinity Chromatography Media, Cytiva
Protein Purification Protein Purification Resins
Cobalt-based chromatography medium designed for purification of histidine-tagged recombinant proteins by immobilized metal affinity chromatography (IMAC)
  • Suitable when Ni²⁺ is not the optimal choice of metal ion
  • Compatible with commonly used IMAC reagents
  • Allows protein purification under native or denaturing conditions
  • Used with prokaryotic and eukaryotic expression systems
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty