Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Ethanol absolute ≥100% (v/v) USP for molecular biology (200 Proof)

Supplier: Decon Labs
Danger
Synonyms: Ethyl alcohol

3916 3916EA
71006-010CS 2555.73 USD InStock
71006-010 71006-012
Ethanol absolute ≥100% (v/v) USP for molecular biology (200 Proof)
Ethanol
Decon’s 200 proof ethanol (100% ethanol) is a Molecular Biology Grade, pure ethanol product made to USP specifications and is tested for DNAase / RNAase activity and its ability to precipitate DNA from solution. Molecular Biology Grade Ethanol can be used in absolute form or diluted for use in most applications in Molecular Biology Laboratories.

Ordering Information: Pure Ethanol products are taxed according to the government schedule of excise taxes detailed in 27 CFR Chapter 1 section 27.41. This excise tax is included in the cost of this product. There is no need for Federal Permits.

  • Total alcohol: 99.5%
  • Ethyl alcohol: 99.5%
  • Denaturant: none


Formula: C₂H₅OH
MW: 46.07 g/mol
Boiling Pt: 78.3 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.7895 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806
Order Now
Specification Test Results
RNA-ase Negative Pass
DNA-ase Negative Pass
Assay 200 proof 99.5 (% v/v) Ethanol
Water 0.1
Specific Gravity @15.56˚C (USP) 0.7962
Specific Gravity @15.0˚C (JP) 0.797
Relative Density @ 20˚C (BP/EP) 0.790-0.793
Color Passes USP test for color
Clarity Passes USP Test for Clarity
Acidity 30 ppm
Alkalinity Passes USP Test
Non Volatile Residue 25 ppm
Sum of Acetal &Acetaldehyde 10 ppm
Benzene 2 ppm
Methanol 100 ppm
UV Absorbance @ 240 nm 0.40 A (USP/BP/EP/JP)
UV Absorbance @ 250nm 0.30 A (USP/BP/EP/JP)
UV Absorbance @ 260nm 0.30 A (USP/BP/EP/JP)
UV Absorbance @ 270 0.10 A (USP/BP/EP/JP)
UV Absorbance @ 235- 340 nm Pass (USP/BP/EP/JP)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty