Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium peroxide ≥93.0% ACS

Supplier: VWR
Danger
Synonyms: di-Sodium peroxide

ANAC84364-300 ANAC84364-340 ANAC84364-580 ANAC84364-300
CA71008-758EA 4342.58 USD InStock
CA71008-758 CA71008-754 CA71008-756 71008-754
Sodium peroxide ≥93.0% ACS
Sodium peroxide


Formula: Na₂O₂
MW: 77.98 g/mol
Melting Pt: 660 °C
Density: 2.805 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003497
CAS Number: 1313-60-6
EINECS: 215-209-4
UN: 1504
ADR: 5.1,I
Merck Index: 12,08800
Order Now
Specification Test Results
Assay (Na2O2) 93.0% min.
Chloride (Cl) 0.002% max.
Phosphate (PO4) 5 ppm max.
Sulfate (SO4) 0.001% max.
Heavy metals (as Pb) 0.002% max.
Iron (Fe) 0.005% max.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty