Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 93-98%, Environmental Grade

Supplier: VWR
Danger

S010301-ANNQ09 ANAC88360-460_U
71008-942CS 773.1 USD InStock
71008-942 CA71008-942
Sulfuric acid 93-98%, Environmental Grade
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Merck Index: 14,08974
Order Now
Specification Test Results
Assay (H2SO4) 93 - 98 %
Colour (APHA) 10 max.
Aluminum (Al) 1 ppb max.
Antimony (Sb) 1 ppb max.
Arsenic (As) 0.5 ppb max.
Barium(Ba) 0.1 ppb max.
Beryllium (Be) 0.1 ppb max.
Bismuth (Bi) 0.1 ppb max.
Cadmium (Cd) 0.5 ppb max.
Calcium (Ca) (20) 1 ppb max.
Cerium (Ce) 0.1 ppb max.
Cesium (Cs) 0.1 ppb max.
Chromium (Cr) 0.5 ppb max.
Cobalt (Co) 0.5 ppb max.
Copper (Cu) 0.5 ppb max.
Dysprosium (Dy) 0.1 ppb max.
Erbium (Er) 0.1 ppb max.
Europium (Eu) 0.1 ppb max.
Gadolinium (Gd) 0.1 ppb max.
Gallium(Ga) 0.1 ppb max.
Germanium(Ge) 1 ppb max.
Gold (Au) 0.5 ppb max.
Hafnium (Hf) 0.1 ppb max.
Holmium (Ho) 0.1 ppb max.
Indium (In) 0.1 ppb max.
Iron (Fe) 1 ppb max.
Lanthanum (La) 0.1 ppb max.
Lead (Pb) 0.1 ppb max.
Lithium (Li) 0.5 ppb max.
Lutetium (Lu) 0.1 ppb max.
Magnesium (Mg) (5) 1 ppb max.
Manganese (Mn) 0.5 ppb max.
Mercury (Hg) 0.1 ppb max.
Molybdenum (Mo) 0.5 ppb max.
Neodymium (Nd) 0.1 ppb max.
Nickel (Ni) 0.5 ppb max.
Niobium (Nb) 0.1 ppb max.
Palladium (Pd) info only
Platinum (Pt) info only
Potassium (K) (30) 1 ppb max.
Praseodymium (Pr) 0.1 ppb max.
Rhodium(Rh) 0.5 ppb max.
Rubidium (Rb) 0.5 ppb max.
Samarium (Sm) 0.1 ppb max.
Scandium (Sc) 0.1 ppb max.
Selenium (Se) 10 ppb max.
Silver (Ag) 1 ppb max.
Sodium (Na) 1 ppb max.
Strontium (Sr) 0.5 ppb max.
Tantalum (Ta) info only
Tellurium (Te) 0.1 ppb max.
Terbium (Tb) 0.1 ppb max.
Thallium (Tl) 0.1 ppb max.
Thorium (Th) 0.1 ppb max.
Thulium (Tm) 0.1 ppb max.
Tin (Sn) 1 ppb max.
Titanium (Ti) 1 ppb max.
Tungsten (W) 0.5 ppb max.
Uranium (U) 0.1 ppb max.
Vanadium (V) 0.5 ppb max.
Ytterbium (Yb) 0.1 ppb max.
Yttrium (Y) 0.1 ppb max.
Zinc (Zn) 1 ppb max.
Zirconium (Zr) 0.5 ppb max.
Maximums stated applicable at time of bottling. Values in brackets are likely maximum values after storage in glass.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty